Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630733.3 | XP_005630790.1 | 4 - 5 | 484 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005630733.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
