Assay Details
| Gene Symbol: AOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119975.2 | XP_013975450.1 | 1986 | 60 | ||
| XM_532759.5 | XP_532759.1 | 1933 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: AOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119975.2 | XP_013975450.1 | 1986 | 60 | ||
| XM_532759.5 | XP_532759.1 | 1933 | 60 | |||
Set Membership: |
|