Assay Details
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616187.3 | XP_005616244.1 | 4 - 5 | 870 | 79 | |
XM_005616188.3 | XP_005616245.1 | 4 - 5 | 873 | 79 | ||
XM_005616189.3 | XP_005616246.1 | 4 - 5 | 748 | 79 | ||
XM_014118494.2 | XP_013973969.1 | 3 - 4 | 731 | 79 | ||
XM_533584.6 | XP_533584.1 | 4 - 5 | 831 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
