Assay Details
Gene Symbol: LGALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284467.1 | NP_001271396.1 | 2 - 3 | 249 | 97 | |
XM_014117247.2 | XP_013972722.1 | 3 - 4 | 339 | 97 | ||
GenBank mRNA | HQ637385.1 | - | 2 - 3 | 249 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
