Assay Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290092.1 | NP_001277021.1 | 1 | 126 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290092.1 transcripts.
Set Membership: |
|

LEPROTL1
Genomic Map