Assay Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290092.1 | NP_001277021.1 | 1 | 394 | 83 | |
| XM_005629907.3 | XP_005629964.1 | 321 | 83 | |||
More Information
Set Membership: |
|

LEPROTL1
Genomic Map