Assay Details
| Gene Symbol: STARD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014112008.2 | XP_013967483.1 | 612 | 69 | ||
| XM_014112016.2 | XP_013967491.1 | 562 | 69 | |||
| XM_022416508.1 | XP_022272216.1 | 726 | 69 | |||
| XM_536287.5 | XP_536287.3 | 726 | 69 | |||
More Information
Set Membership: |
|

STARD4
Genomic Map