Assay Details
| Gene Symbol: MDM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415420.1 | XP_022271128.1 | 4 - 5 | 475 | 84 | |
| XM_022415421.1 | XP_022271129.1 | 4 - 5 | 540 | 84 | ||
| XM_022415422.1 | XP_022271130.1 | 4 - 5 | 517 | 84 | ||
| XM_022415423.1 | XP_022271131.1 | 1 - 2 | 55 | 84 | ||
| XM_022415424.1 | XP_022271132.1 | 1 - 2 | 82 | 84 | ||
| XM_022415425.1 | XP_022271133.1 | 4 - 5 | 598 | 84 | ||
| XM_022415426.1 | XP_022271134.1 | 4 - 5 | 592 | 84 | ||
| XM_536098.6 | XP_536098.3 | 4 - 5 | 567 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022415423.1, XM_022415424.1 transcripts.
Set Membership: |
|

MDM4
Genomic Map