Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623328.3 | XP_005623385.1 | 5 - 6 | 676 | 109 | |
| XM_022421922.1 | XP_022277630.1 | 5 - 6 | 515 | 109 | ||
| XM_537414.6 | XP_537414.2 | 5 - 6 | 676 | 109 | ||
More Information
Set Membership: |
|

BRMS1L
Genomic Map