Assay Details
Gene Symbol: CCSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014112932.2 | XP_013968407.1 | 6 - 7 | 2152 | 87 | |
XM_014112935.2 | XP_013968410.1 | 7 - 8 | 2176 | 87 | ||
XM_014112936.2 | XP_013968411.1 | 7 - 8 | 2176 | 87 | ||
XM_014112937.2 | XP_013968412.1 | 7 - 8 | 2176 | 87 | ||
XM_014112938.2 | XP_013968413.1 | 7 - 8 | 2176 | 87 | ||
XM_014112939.2 | XP_013968414.1 | 7 - 8 | 2175 | 87 | ||
XM_014112940.2 | XP_013968415.1 | 7 - 8 | 2176 | 87 | ||
XM_014112942.2 | XP_013968417.1 | 4 - 5 | 509 | 87 | ||
XM_014112943.2 | XP_013968418.1 | 3 - 4 | 510 | 87 | ||
XM_022417611.1 | XP_022273319.1 | 6 - 7 | 2114 | 87 | ||
XM_022417612.1 | XP_022273320.1 | 7 - 8 | 2176 | 87 | ||
XM_022417614.1 | XP_022273322.1 | 7 - 8 | 2175 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
