Assay Details
Gene Symbol: NRBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536362.5 | XP_536362.3 | 1 - 2 | 233 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_536362.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
