Assay Details
| Gene Symbol: PNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423056.1 | XP_022278764.1 | 286 | 63 | ||
| XM_537661.6 | XP_537661.2 | 282 | 63 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423056.1 | XP_022278764.1 | 286 | 63 | ||
| XM_537661.6 | XP_537661.2 | 282 | 63 | |||
Set Membership: |
|