Assay Details
| Gene Symbol: GGPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638980.4 | XP_003639028.1 | 695 | 97 | ||
| XM_005618806.2 | XP_005618863.1 | 390 | 97 | |||
| XM_022417400.1 | XP_022273108.1 | 726 | 97 | |||
| XM_022417401.1 | XP_022273109.1 | 405 | 97 | |||
| XM_022417402.1 | XP_022273110.1 | 246 | 97 | |||
| XM_536340.7 | XP_536340.2 | 580 | 97 | |||
More Information
Set Membership: |
|

GGPS1
Genomic Map