Assay Details
| Gene Symbol: C9H9orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537817.6 | XP_537817.1 | 1 - 2 | 170 | 70 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length less than or equal to 70  Detect Genomic DNA  Canis familiaris  Probe spans exons | 
 Back To Top
 Back To Top
 C9H9orf78
C9H9orf78 Genomic Map
Genomic Map Back To Top
 Back To Top