Assay Details
| Gene Symbol: C9H9orf78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537817.6 | XP_537817.1 | 6 - 7 | 624 | 92 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C9H9orf78
Genomic Map