Assay Details
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621502.3 | XP_005621559.1 | 3 - 4 | 430 | 72 | |
XM_536955.6 | XP_536955.2 | 3 - 4 | 453 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
