Assay Details
| Gene Symbol: C5H17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619942.3 | XP_005619999.1 | 1 - 2 | 101 | 51 | |
| XM_005619944.3 | XP_005620001.1 | 1 - 2 | 102 | 51 | ||
| XM_005619946.3 | XP_005620003.1 | 1 - 2 | 119 | 51 | ||
| XM_022418473.1 | XP_022274181.1 | 1 - 2 | 103 | 51 | ||
| XM_022418474.1 | XP_022274182.1 | 1 - 2 | 104 | 51 | ||
| XM_536614.6 | XP_536614.2 | 1 - 2 | 102 | 51 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005619942.3, XM_005619944.3, XM_005619946.3, XM_022418473.1, XM_022418474.1, XM_536614.6 transcripts.
Set Membership: |
|

C5H17orf49
Genomic Map