Assay Details
| Gene Symbol: C5H17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619942.3 | XP_005619999.1 | 5 - 6 | 591 | 80 | |
| XM_005619946.3 | XP_005620003.1 | 4 - 5 | 507 | 80 | ||
More Information
Set Membership: |
|

C5H17orf49
Genomic Map