Assay Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434301.4 | XP_003434349.1 | 2 - 3 | 274 | 151 | |
| XM_005640811.3 | XP_005640868.1 | 2 - 3 | 185 | 151 | ||
| XM_022415449.1 | XP_022271157.1 | 2 - 3 | 76 | 151 | ||
| XM_536114.6 | XP_536114.3 | 2 - 3 | 325 | 151 | ||
More Information
Set Membership: |
|

GLRX2
Genomic Map