Assay Details
Gene Symbol: LOC479708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620930.3 | XP_005620987.1 | 1405 | 61 | ||
XM_022419526.1 | XP_022275234.1 | 1406 | 61 | |||
XM_022419527.1 | XP_022275235.1 | 1406 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
