Assay Details
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621787.3 | XP_005621844.1 | 8 - 9 | 1030 | 75 | |
XM_005621788.3 | XP_005621845.1 | 7 - 8 | 960 | 75 | ||
XM_005621789.3 | XP_005621846.1 | 7 - 8 | 945 | 75 | ||
XM_005621790.3 | XP_005621847.1 | 6 - 7 | 875 | 75 | ||
XM_022420339.1 | XP_022276047.1 | 4 - 5 | 526 | 75 | ||
XM_537031.6 | XP_537031.2 | 8 - 9 | 1027 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
