Assay Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014118117.2 | XP_013973592.1 | 2 - 3 | 503 | 73 | |
XM_022426269.1 | XP_022281977.1 | 4 - 5 | 610 | 73 | ||
XM_022426270.1 | XP_022281978.1 | 4 - 5 | 610 | 73 | ||
XM_539013.6 | XP_539013.3 | 3 - 4 | 563 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
