Assay Details
Gene Symbol: CYB561A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631673.3 | XP_005631730.1 | 6 - 7 | 718 | 63 | |
XM_005631674.3 | XP_005631731.1 | 5 - 6 | 658 | 63 | ||
XM_005631676.3 | XP_005631733.1 | 4 - 5 | 773 | 63 | ||
XM_005631677.3 | XP_005631734.1 | 4 - 5 | 634 | 63 | ||
XM_014121268.2 | XP_013976743.1 | 5 - 6 | 825 | 63 | ||
XM_014121270.2 | XP_013976745.1 | 5 - 6 | 726 | 63 | ||
XM_022405822.1 | XP_022261530.1 | 6 - 7 | 752 | 63 | ||
XM_022405823.1 | XP_022261531.1 | 5 - 6 | 592 | 63 | ||
XM_022405824.1 | XP_022261532.1 | 6 - 7 | 805 | 63 | ||
XM_022405825.1 | XP_022261533.1 | 6 - 7 | 763 | 63 | ||
XM_022405826.1 | XP_022261534.1 | 6 - 7 | 771 | 63 | ||
XM_022405827.1 | XP_022261535.1 | 5 - 6 | 700 | 63 | ||
XM_022405828.1 | XP_022261536.1 | 5 - 6 | 666 | 63 | ||
XM_540920.6 | XP_540920.2 | 6 - 7 | 930 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
