Assay Details
| Gene Symbol: TRIM80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422734.1 | XP_022278442.1 | 160 | 109 | ||
| XM_540426.6 | XP_540426.3 | 160 | 109 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TRIM80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422734.1 | XP_022278442.1 | 160 | 109 | ||
| XM_540426.6 | XP_540426.3 | 160 | 109 | |||
Set Membership: |
|