Assay Details
| Gene Symbol: MAP3K11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_540853.6 | XP_540853.5 | 4 - 5 | 1874 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP3K11| Gene Symbol: MAP3K11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_540853.6 | XP_540853.5 | 4 - 5 | 1874 | 67 | |
Set Membership: |
|