Assay Details
Gene Symbol: FIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638798.4 | XP_003638846.1 | 2 - 3 | 499 | 77 | |
XM_005616109.2 | XP_005616166.1 | 2 - 3 | 601 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
