Assay Details
| Gene Symbol: DPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631484.3 | XP_005631541.1 | 4 - 5 | 595 | 69 | |
| XM_861495.5 | XP_866588.1 | 4 - 5 | 595 | 69 | ||
More Information
Set Membership: |
|

DPF2
Genomic Map