Assay Details
| Gene Symbol: GALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630359.3 | XP_005630416.1 | 2 - 3 | 332 | 70 | |
| XM_540154.6 | XP_540154.2 | 2 - 3 | 543 | 70 | ||
More Information
Set Membership: |
|

GALM
Genomic Map