Assay Details
Gene Symbol: SGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314125.1 | NP_001301054.1 | 4 - 5 | 390 | 58 | |
XM_005633093.2 | XP_005633150.1 | 5 - 6 | 565 | 58 | ||
XM_005633094.2 | XP_005633151.1 | 5 - 6 | 565 | 58 | ||
XM_005633095.3 | XP_005633152.1 | 5 - 6 | 685 | 58 | ||
XM_005633097.3 | XP_005633154.1 | 5 - 6 | 453 | 58 | ||
XM_005633098.3 | XP_005633155.1 | 5 - 6 | 557 | 58 | ||
XM_022406249.1 | XP_022261957.1 | 5 - 6 | 757 | 58 | ||
XM_022406250.1 | XP_022261958.1 | 5 - 6 | 517 | 58 | ||
GenBank mRNA | LC021529.1 | - | 4 - 5 | 390 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
