Assay Details
| Gene Symbol: POP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627813.3 | XP_005627870.1 | 262 | 59 | ||
| XM_005627814.3 | XP_005627871.1 | 280 | 59 | |||
| XM_539099.6 | XP_539099.3 | 262 | 59 | |||
More Information
Set Membership: |
|

POP1
Genomic Map