Assay Details
Gene Symbol: APLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626176.3 | XP_005626233.1 | 5 - 6 | 649 | 121 | |
XM_022424830.1 | XP_022280538.1 | 6 - 7 | 810 | 121 | ||
XM_022424831.1 | XP_022280539.1 | 6 - 7 | 825 | 121 | ||
XM_022424832.1 | XP_022280540.1 | 6 - 7 | 825 | 121 | ||
XM_538525.6 | XP_538525.2 | 6 - 7 | 825 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
