Assay Details
| Gene Symbol: ASRGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631627.3 | XP_005631684.1 | 3 - 4 | 604 | 64 | |
| XM_005631628.3 | XP_005631685.1 | 3 - 4 | 616 | 64 | ||
| XM_540910.6 | XP_540910.2 | 3 - 4 | 549 | 64 | ||
More Information
Set Membership: |
|

ASRGL1
Genomic Map