Assay Details
Gene Symbol: FUCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615687.2 | XP_005615744.1 | 1363 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
