Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424448.1 | XP_022280156.1 | 2 - 3 | 180 | 76 | |
| XM_022424449.1 | XP_022280157.1 | 2 - 3 | 269 | 76 | ||
| XM_022424450.1 | XP_022280158.1 | 2 - 3 | 416 | 76 | ||
| XM_022424451.1 | XP_022280159.1 | 2 - 3 | 1303 | 76 | ||
| XM_022424452.1 | XP_022280160.1 | 2 - 3 | 133 | 76 | ||
| XM_022424453.1 | XP_022280161.1 | 2 - 3 | 3581 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022424449.1, XM_022424450.1, XM_022424451.1, XM_022424452.1, XM_022424453.1 transcripts.
Set Membership: |
|

MAFF
Genomic Map