Assay Details
| Gene Symbol: LOC100687510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413430.1 | XP_022269138.1 | 3 | 497 | 79 | |
| Gene Symbol: SNRPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427684.1 | XP_022283392.1 | 4 - 5 | 746 | 79 | |
More Information
Set Membership: |
|

LOC100687510
Genomic Map