Assay Details
Gene Symbol: ACP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634423.3 | XP_005634480.1 | 4 - 5 | 685 | 68 | |
XM_005634424.3 | XP_005634481.1 | 4 - 5 | 692 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
