Assay Details
Gene Symbol: LOC486353 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_543479.5 | XP_543479.2 | 1 - 2 | 115 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_543479.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
