Assay Details
| Gene Symbol: MFSD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618506.3 | XP_005618563.1 | 11 - 12 | 1309 | 58 | |
| XM_022417111.1 | XP_022272819.1 | 10 - 11 | 1105 | 58 | ||
More Information
Set Membership: |
|

MFSD10
Genomic Map