Assay Details
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640194.3 | XP_005640251.1 | 4 - 5 | 722 | 74 | |
XM_022414682.1 | XP_022270390.1 | 4 - 5 | 486 | 74 | ||
XM_545482.6 | XP_545482.2 | 4 - 5 | 462 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
