Assay Details
| Gene Symbol: RAB5IF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433257.4 | XP_003433305.1 | 1 - 2 | 345 | 93 | |
| XM_542982.6 | XP_542982.2 | 1 - 2 | 335 | 93 | ||
More Information
Set Membership: |
|

RAB5IF
Genomic Map