Assay Details
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284456.1 | NP_001271385.1 | 1 - 2 | 240 | 79 | |
| GenBank mRNA | AB648939.1 | - | 1 - 2 | 240 | 79 | |
More Information
Set Membership: |
|

CX3CL1
Genomic Map