Assay Details
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122609.1 | NP_001116081.1 | 2 - 3 | 207 | 85 | |
XM_005634693.3 | XP_005634750.1 | 3 - 4 | 376 | 85 | ||
XM_005634694.3 | XP_005634751.1 | 2 - 3 | 1880 | 85 | ||
XM_005634696.3 | XP_005634753.1 | 2 - 3 | 525 | 85 | ||
XM_014106836.2 | XP_013962311.1 | 2 - 3 | 1923 | 85 | ||
XM_014106837.2 | XP_013962312.1 | 3 - 4 | 333 | 85 | ||
XM_022408815.1 | XP_022264523.1 | 3 - 4 | 723 | 85 | ||
XM_022408816.1 | XP_022264524.1 | 2 - 3 | 653 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
