Assay Details
| Gene Symbol: MUL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413821.1 | XP_022269529.1 | 3 - 4 | 232 | 65 | |
| XM_544520.5 | XP_544520.2 | 3 - 4 | 432 | 65 | ||
More Information
Set Membership: |
|

MUL1
Genomic Map