Assay Details
| Gene Symbol: LOC487628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109562.2 | XP_013965037.1 | 8 - 9 | 1396 | 106 | |
| XM_022412902.1 | XP_022268610.1 | 8 - 9 | 1379 | 106 | ||
More Information
Set Membership: |
|

LOC487628
Genomic Map