Assay Details
| Gene Symbol: ZDHHC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635550.3 | XP_005635607.1 | 409 | 118 | ||
| XM_005635551.3 | XP_005635608.1 | 399 | 118 | |||
| XM_014107410.2 | XP_013962885.1 | 402 | 118 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZDHHC20
Genomic Map