Assay Details
| Gene Symbol: CNOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619263.3 | XP_005619320.1 | 3 - 4 | 552 | 141 | |
| XM_022417770.1 | XP_022273478.1 | 3 - 4 | 517 | 141 | ||
| XM_022417771.1 | XP_022273479.1 | 2 - 3 | 320 | 141 | ||
| XM_022417772.1 | XP_022273480.1 | 3 - 4 | 636 | 141 | ||
| XM_546280.6 | XP_546280.1 | 3 - 4 | 634 | 141 | ||
More Information
Set Membership: |
|

CNOT8
Genomic Map