Assay Details
Gene Symbol: CST8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634704.3 | XP_005634761.1 | 2 - 3 | 616 | 124 | |
XM_022409276.1 | XP_022264984.1 | 2 - 3 | 621 | 124 | ||
XM_022409277.1 | XP_022264985.1 | 2 - 3 | 512 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
