Assay Details
Gene Symbol: NPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014109543.2 | XP_013965018.1 | 4 - 5 | 751 | 56 | |
XM_022412963.1 | XP_022268671.1 | 4 - 5 | 751 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
