Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424081.1 | XP_022279789.1 | 1892 | 84 | ||
| XM_022424082.1 | XP_022279790.1 | 1466 | 84 | |||
| XM_843974.4 | XP_849067.2 | 1177 | 84 | |||
More Information
Set Membership: |
|

NEMP1
Genomic Map