Assay Details
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014113508.2 | XP_013968983.1 | 1 - 2 | 194 | 70 | |
XM_844226.4 | XP_849319.2 | 1 - 2 | 188 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
